Assay Details
| Gene Symbol: Ap4m1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021392.4 | NP_067367.3 | 10 - 11 | 1005 | 117 | |
| GenBank mRNA | AF242858.1 | - | 10 - 11 | 873 | 117 | |
| AK031638.1 | - | 10 - 11 | 1008 | 117 | ||
| BC011174.1 | - | 10 - 11 | 890 | 4207033 | 117 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ap4m1
Genomic Map