Assay Details
| Gene Symbol: Ap4m1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021392.4 | NP_067367.3 | 7 - 8 | 780 | 65 | |
| GenBank mRNA | AF242858.1 | - | 7 - 8 | 648 | 65 | |
| AK031638.1 | - | 7 - 8 | 783 | 65 | ||
| BC011174.1 | - | 7 - 8 | 665 | 4207033 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ap4m1
Genomic Map