Assay Details
| Gene Symbol: Zscan21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044704.2 | NP_001038169.1 | 2 - 3 | 163 | 72 | |
| NM_011757.3 | NP_035887.2 | 2 - 3 | 182 | 72 | ||
| GenBank mRNA | AK146776.1 | - | 2 - 3 | 156 | 72 | |
| AK146875.1 | - | 2 - 3 | 143 | 72 | ||
| CF915830.1 | - | 2 - 3 | 107 | 72 | ||
| DV651309.1 | - | 2 - 3 | 195 | 72 | ||
| U41671.1 | - | 2 - 3 | 113 | 72 | ||
| X63747.1 | - | 1 - 2 | 873 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK146875.1, NM_001044704.2, NM_011757.3, U41671.1, X63747.1 transcripts.
Set Membership: |
|

Zscan21
Genomic Map