Assay Details
| Gene Symbol: Gtf2i | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080746.2 | NP_001074215.1 | 31 - 32 | 3232 | 90 | |
| NM_001080747.2 | NP_001074216.1 | 30 - 31 | 3169 | 90 | ||
| NM_001080748.2 | NP_001074217.1 | 29 - 30 | 3112 | 90 | ||
| NM_001080749.2 | NP_001074218.1 | 29 - 30 | 3124 | 90 | ||
| NM_010365.4 | NP_034495.2 | 30 - 31 | 3175 | 90 | ||
| GenBank mRNA | AF017085.1 | - | 31 - 32 | 2876 | 90 | |
| AF043219.1 | - | 29 - 30 | 90 | |||
| AF043220.1 | - | 28 - 29 | 90 | |||
| AK013348.1 | - | 8 - 9 | 709 | 90 | ||
| AK145698.1 | - | 21 - 22 | 2060 | 90 | ||
| AK147201.1 | - | 29 - 30 | 3082 | 90 | ||
| AY030290.1 | - | 30 - 31 | 2862 | 90 | ||
| AY030291.1 | - | 30 - 31 | 2868 | 90 | ||
| AY030292.1 | - | 29 - 30 | 2805 | 90 | ||
| AY030293.1 | - | 31 - 32 | 2925 | 90 | ||
| BC047387.1 | - | 29 - 30 | 2771 | 90 | ||
| BC053044.1 | - | 31 - 32 | 3201 | 5704935 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gtf2i
Genomic Map