Assay Details
| Gene Symbol: Lmtk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081109.1 | NP_001074578.1 | 7 - 8 | 1027 | 118 | |
| GenBank mRNA | AB288872.1 | - | 7 - 8 | 855 | 118 | |
| AK158724.1 | - | 7 - 8 | 1028 | 118 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lmtk2
Genomic Map