Assay Details
| Gene Symbol: Tsg101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021884.3 | NP_068684.1 | 4 - 5 | 427 | 109 | |
| GenBank mRNA | AK014049.1 | - | 4 - 5 | 385 | 109 | |
| AK146229.1 | - | 4 - 5 | 437 | 109 | ||
| AK146602.1 | - | 4 - 5 | 428 | 109 | ||
| AK150369.1 | - | 4 - 5 | 466 | 109 | ||
| AK179695.1 | - | 4 - 5 | 346 | 109 | ||
| AK185554.1 | - | 4 - 5 | 342 | 109 | ||
| AK207014.1 | - | 3 - 4 | 309 | 109 | ||
| AK216304.1 | - | 3 - 4 | 311 | 109 | ||
| BC005424.1 | - | 4 - 5 | 443 | 3490301 | 109 | |
| BC085308.1 | - | 4 - 5 | 447 | 6395694 | 109 | |
| U52945.1 | - | 4 - 5 | 391 | 109 | ||
More Information
Set Membership: |
|

Tsg101
Genomic Map