Assay Details
| Gene Symbol: Csf2rb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007780.4 | NP_031806.3 | 11 - 12 | 1676 | 82 | |
| GenBank mRNA | AF483496.1 | - | 10 - 11 | 1418 | 82 | |
| AF483497.1 | - | 10 - 11 | 1418 | 82 | ||
| AK133420.1 | - | 1620 | 82 | |||
| AK152608.1 | - | 11 - 12 | 1616 | 82 | ||
| AK154286.1 | - | 11 - 12 | 1613 | 82 | ||
| AK155109.1 | - | 11 - 12 | 1647 | 82 | ||
| AK155215.1 | - | 11 - 12 | 1556 | 82 | ||
| AK155699.1 | - | 11 - 12 | 1613 | 82 | ||
| AK155803.1 | - | 11 - 12 | 1613 | 82 | ||
| AK155885.1 | - | 11 - 12 | 1617 | 82 | ||
| AK171477.1 | - | 11 - 12 | 1613 | 82 | ||
| BC113202.1 | - | 12 - 13 | 1687 | 40048156 | 82 | |
| M34397.1 | - | 12 - 13 | 1705 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P123635 | BC113202.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P123635 | M34397.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P123635 | AK155699.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P123635 | AK155109.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P123635 | AK152608.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P52618 | AK154286.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P52618 | AI467222.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P52618 | NM_007780.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P52618 | AK154286.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P52618 | AI467222.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P52618 | NM_007780.4 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 726870 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 256694 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 528893 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 939358 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 105851 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 327691 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 748586 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 251734 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 787631 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 247963 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 874236 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 122380 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 43271 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 103876 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P100465 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P141985 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P52618 | AK154286.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P52618 | AI467222.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P52618 | NM_007780.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P52618 | AK154286.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P52618 | AI467222.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P52618 | NM_007780.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P123635 | BC113202.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P123635 | M34397.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P123635 | AK155699.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P123635 | AK155109.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P123635 | AK152608.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P492939 | BC113202.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P492939 | M34397.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P492939 | AK171477.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P492939 | AK155822.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P492939 | AK155885.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P664853 | BC113202.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P664853 | M34397.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P664853 | S78451.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P664853 | AK171477.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P664853 | AK155822.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P52618 | AK154286.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P52618 | AI467222.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P52618 | NM_007780.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P52618 | AK154286.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P52618 | AI467222.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P52618 | NM_007780.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P664853 | BC113202.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P664853 | M34397.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P664853 | S78451.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P664853 | AK171477.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P664853 | AK155822.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P492939 | BC113202.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P492939 | M34397.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P492939 | AK171477.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P492939 | AK155822.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P492939 | AK155885.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123635 | BC113202.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123635 | M34397.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123635 | AK155699.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123635 | AK155109.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123635 | AK152608.1 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12374531 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12374532 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12374536 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11944728 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10425067 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10425070 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10425075 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10425072 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10425069 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10425073 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10425071 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10425068 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10425074 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455660_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421326_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10425066 | - | - |
| Mouse Genome 430 2.0 Array | 1450200_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1455660_at | - | - |
| Mouse Genome 430 2.0 Array | 1455660_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1450200_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1421326_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1421326_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12983_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12983_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500004671.mm.1 | NM_007780.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500004671.mm.1 | NM_007780.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500004671.mm.1 | TC1500000641.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500004675.mm.1 | NM_007780.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500004675.mm.1 | NM_007780.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500004675.mm.1 | TC1500000641.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500004667.mm.1 | NM_007780.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500004667.mm.1 | NM_007780.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500004667.mm.1 | TC1500000641.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500004674.mm.1 | NM_007780.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500004674.mm.1 | NM_007780.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500004674.mm.1 | TC1500000641.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500004672.mm.1 | NM_007780.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500004672.mm.1 | NM_007780.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500004672.mm.1 | TC1500000641.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500004670.mm.1 | NM_007780.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500004670.mm.1 | NM_007780.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500004670.mm.1 | TC1500000641.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500004673.mm.1 | NM_007780.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500004673.mm.1 | NM_007780.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500004673.mm.1 | TC1500000641.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500004665.mm.1 | NM_007780.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500004665.mm.1 | NM_007780.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500004665.mm.1 | TC1500000641.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500004669.mm.1 | NM_007780.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500004669.mm.1 | NM_007780.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500004669.mm.1 | TC1500000641.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500004666.mm.1 | NM_007780.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500004666.mm.1 | NM_007780.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500004666.mm.1 | TC1500000641.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1421326_PM_at | - | - |
| Mouse Expression Set 430 | 1421326_at | - | - |
| Mouse Expression Set 430 | 1450200_PM_s_at | - | - |
| Mouse Expression Set 430 | 1450200_s_at | - | - |
| Mouse Expression Set 430 | 1455660_PM_at | - | - |
| Mouse Expression Set 430 | 1455660_at | - | - |
| Murine Genome U74v2 Set | 94747_PM_at | - | - |
| Murine Genome U74v2 Set | 94748_g_at | - | - |
| Murine Genome U74v2 Set | 94748_PM_g_at | - | - |
| Murine Genome U74v2 Set | 94747_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421326_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421326_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450200_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450200_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421326_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421326_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455660_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455660_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4480156 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4480369 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6760273 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4480156 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006760273 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004480156 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004480369 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1236070 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1253312 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2688262 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1253312 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2688262 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1236070 | - | - |
| mouse-6 v1.1 (Illumina) | scl012983.8_64-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0012983.1_70-S | - | - |
| mouse-6 v1.1 (Illumina) | scl012983.13_141-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4480156 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6760273 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4480369 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl012983.8_64-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl012983.13_141-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0012983.1_70-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4480369 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4480156 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6760273 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1236070 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2688262 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1253312 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1253312 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1236070 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2688262 | - | - |

Csf2rb
Genomic Map