Assay Details
| Gene Symbol: Rfc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020022.2 | NP_064406.1 | 2 - 3 | 269 | 113 | |
| GenBank mRNA | AK011353.1 | - | 2 - 3 | 185 | 113 | |
| AK011649.1 | - | 2 - 3 | 183 | 113 | ||
| AK075997.1 | - | 2 - 3 | 181 | 113 | ||
| AK154681.1 | - | 2 - 3 | 223 | 113 | ||
| BC023028.1 | - | 2 - 3 | 190 | 5374851 | 113 | |
| BC099370.1 | - | 2 - 3 | 233 | 30937690 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rfc2
Genomic Map