Assay Details
| Gene Symbol: Pla2g4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168504.1 | NP_001161976.1 | 1 - 2 | 112 | 75 | |
| GenBank mRNA | BC117808.1 | - | 1 - 2 | 112 | 40092567 | 75 |
| BC117809.1 | - | 1 - 2 | 112 | 40092569 | 75 | |
| BC129974.1 | - | 1 - 2 | 112 | 40092571 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pla2g4c
Genomic Map