Assay Details
| Gene Symbol: Slk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164639.1 | NP_001158111.1 | 6 - 7 | 958 | 109 | |
| NM_009289.3 | NP_033315.2 | 6 - 7 | 958 | 109 | ||
| GenBank mRNA | AF039574.1 | - | 6 - 7 | 963 | 109 | |
| AF112855.1 | - | 6 - 7 | 847 | 109 | ||
| AK029491.1 | - | 6 - 7 | 825 | 109 | ||
| AK122218.1 | - | 6 - 7 | 1299 | 109 | ||
| BC131674.1 | - | 6 - 7 | 859 | 40110915 | 109 | |
| BC131675.1 | - | 6 - 7 | 859 | 40110921 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slk
Genomic Map