Assay Details
| Gene Symbol: Slk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164639.1 | NP_001158111.1 | 8 - 9 | 1172 | 60 | |
| NM_009289.3 | NP_033315.2 | 8 - 9 | 1172 | 60 | ||
| GenBank mRNA | AF039574.1 | - | 8 - 9 | 1177 | 60 | |
| AF112855.1 | - | 8 - 9 | 1061 | 60 | ||
| AK029491.1 | - | 8 - 9 | 1039 | 60 | ||
| AK122218.1 | - | 8 - 9 | 1513 | 60 | ||
| BC131674.1 | - | 8 - 9 | 1073 | 40110915 | 60 | |
| BC131675.1 | - | 8 - 9 | 1073 | 40110921 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slk
Genomic Map