Assay Details
| Gene Symbol: Slk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164639.1 | NP_001158111.1 | 9 - 10 | 2520 | 81 | |
| NM_009289.3 | NP_033315.2 | 9 - 10 | 2520 | 81 | ||
| GenBank mRNA | AF039574.1 | - | 9 - 10 | 2525 | 81 | |
| AF112855.1 | - | 9 - 10 | 2409 | 81 | ||
| AK037798.1 | - | 1 - 2 | 494 | 81 | ||
| AK122218.1 | - | 9 - 10 | 2861 | 81 | ||
| BC131674.1 | - | 9 - 10 | 2421 | 40110915 | 81 | |
| BC131675.1 | - | 9 - 10 | 2421 | 40110921 | 81 | |
| CF748583.1 | - | 1 - 2 | 368 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slk
Genomic Map