Assay Details
| Gene Symbol: Ift27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025931.2 | NP_080207.1 | 1 - 2 | 342 | 62 | |
| GenBank mRNA | AK011196.1 | - | 1 - 2 | 340 | 62 | |
| BC017514.1 | - | 1 - 2 | 342 | 4502942 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ift27
Genomic Map