Assay Details
| Gene Symbol: Eif3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018749.2 | NP_061219.2 | 4 - 5 | 459 | 63 | |
| GenBank mRNA | AB012580.1 | - | 4 - 5 | 403 | 63 | |
| AK045228.1 | - | 4 - 5 | 401 | 63 | ||
| AK049767.1 | - | 4 - 5 | 398 | 63 | ||
| AK145689.1 | - | 4 - 5 | 414 | 63 | ||
| AK158344.1 | - | 2 - 3 | 1405 | 63 | ||
| AK168256.1 | - | 4 - 5 | 449 | 63 | ||
| BC018598.1 | - | 4 - 5 | 397 | 63 | ||
| BC089020.1 | - | 4 - 5 | 393 | 30551991 | 63 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Eif3d
Genomic Map