| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500024667.mm.1 |
TC0500002805.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500024667.mm.1 |
NM_133738.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500024667.mm.1 |
NM_133738.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500024665.mm.1 |
TC0500002805.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500024665.mm.1 |
NM_133738.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500024665.mm.1 |
NM_133738.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500024666.mm.1 |
TC0500002805.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500024666.mm.1 |
NM_133738.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500024666.mm.1 |
NM_133738.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500024664.mm.1 |
TC0500002805.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500024664.mm.1 |
NM_133738.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500024664.mm.1 |
NM_133738.1 |
Refseq |