Assay Details
| Gene Symbol: Tnks2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163635.1 | NP_001157107.1 | 17 - 18 | 2470 | 91 | |
| GenBank mRNA | AK149368.1 | - | 3 - 4 | 435 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tnks2
Genomic Map