| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1900004652.mm.1 |
NM_001163635.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004652.mm.1 |
NM_001163635.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004652.mm.1 |
TC1900000521.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900004656.mm.1 |
NM_001163635.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004656.mm.1 |
NM_001163635.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004656.mm.1 |
TC1900000521.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900004650.mm.1 |
NM_001163635.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004650.mm.1 |
NM_001163635.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004650.mm.1 |
TC1900000521.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900004655.mm.1 |
NM_001163635.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004655.mm.1 |
NM_001163635.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004655.mm.1 |
TC1900000521.mm.1 |
MTA TC |