| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1900004620.mm.1 |
NM_001163635.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004620.mm.1 |
NM_001163635.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004620.mm.1 |
TC1900000521.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900004618.mm.1 |
NM_001163635.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004618.mm.1 |
NM_001163635.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004618.mm.1 |
TC1900000521.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900004619.mm.1 |
NM_001163635.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004619.mm.1 |
NM_001163635.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004619.mm.1 |
TC1900000521.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900004621.mm.1 |
NM_001163635.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004621.mm.1 |
NM_001163635.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004621.mm.1 |
TC1900000521.mm.1 |
MTA TC |