Assay Details
| Gene Symbol: Tnks2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163635.1 | NP_001157107.1 | 25 - 26 | 3588 | 77 | |
| GenBank mRNA | AK149368.1 | - | 11 - 12 | 1553 | 77 | |
| BC063101.1 | - | 8 - 9 | 1008 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tnks2
Genomic Map