Assay Details
| Gene Symbol: Tnks2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163635.1 | NP_001157107.1 | 11 - 12 | 1575 | 105 | |
| GenBank mRNA | AK047094.1 | - | 11 - 12 | 1434 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tnks2
Genomic Map