Assay Details
| Gene Symbol: Dpcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172639.2 | NP_766227.1 | 3 - 4 | 300 | 73 | |
| GenBank mRNA | AK077347.1 | - | 3 - 4 | 301 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dpcd
Genomic Map