Assay Details
| Gene Symbol: Cldn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009903.2 | NP_034033.1 | 1 | 1757 | 89 | |
| GenBank mRNA | AB000713.1 | - | 1 | 88 | ||
| AK145152.1 | - | 1 | 1745 | 89 | ||
| BC032172.1 | - | 1 | 1745 | 89 | ||
| BQ031935.1 | - | 1 | 89 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24113992 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24113991 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24113990 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2183453 | BC132376.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2183453 | BC132378.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2183453 | AK162721.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2183453 | BC032172.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2183453 | AF087822.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P356467 | AK145152.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P356467 | BC032172.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P356467 | NM_009903.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P356467 | AB000713.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P356467 | AK145152.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P356467 | BC032172.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P356467 | NM_009903.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P356467 | AB000713.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 593459 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P152063 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2183453 | BC132376.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2183453 | BC132378.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2183453 | AK162721.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2183453 | BC032172.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2183453 | AF087822.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P356467 | AK145152.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P356467 | BC032172.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P356467 | NM_009903.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P356467 | AB000713.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P356467 | AK145152.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P356467 | BC032172.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P356467 | NM_009903.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P356467 | AB000713.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P356467 | AK145152.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P356467 | BC032172.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P356467 | NM_009903.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P356467 | AB000713.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P356467 | AK145152.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P356467 | BC032172.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P356467 | NM_009903.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P356467 | AB000713.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114017 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24113998 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24113980 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24113999 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114024 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114020 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24113993 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24113976 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114001 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114002 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24113986 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24113995 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24113987 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114007 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24113979 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24113984 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114010 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114027 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24113982 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114021 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24113977 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114014 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24113992 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114016 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24113990 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24113988 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114008 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114009 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24113985 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114005 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114004 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114026 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114012 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24113994 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114023 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P152063 | - | - |
| Whole Mouse Genome G4122A | A_51_P356467 | AK145152.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P356467 | BC032172.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P356467 | NM_009903.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P356467 | AB000713.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P356467 | AK145152.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P356467 | BC032172.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P356467 | NM_009903.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P356467 | AB000713.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123756 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P152063 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04772559 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04772547 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04772548 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04197661 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04772556 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04772563 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04772573 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24113991 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24113990 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24113992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24113952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24113998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24113995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24113987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24113988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24113993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24113953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24113955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24113979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24113985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24113950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24113946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24113980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24113959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24113984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24113947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24113999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24113990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24113992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24113949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24113954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24113982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24113977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114030 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24113987 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24113993 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114012 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24113981 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24113979 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114013 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114024 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24113982 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114029 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04106189 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114032 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114036 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24113988 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04106195 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24113977 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114005 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04772548 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114034 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114018 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114030 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24113985 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24113990 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24113994 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114014 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04726603 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04772573 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114015 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04309682 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114033 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114038 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114007 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114002 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114023 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24113995 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114008 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24113980 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114004 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114020 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114001 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114016 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114010 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04772576 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10534397 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534398 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534396 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418283_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10534395 | - | - |
| Mouse Genome 430 2.0 Array | 1418283_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1418283_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12740_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12740_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500030555.mm.1 | TC0500003384.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500030555.mm.1 | NM_009903.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500030555.mm.1 | NM_009903.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500030552.mm.1 | TC0500003384.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500030552.mm.1 | NM_009903.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500030552.mm.1 | NM_009903.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500030554.mm.1 | TC0500003384.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500030554.mm.1 | NM_009903.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500030554.mm.1 | NM_009903.2 | Refseq |
| Mouse Expression Set 430 | 1418283_at | - | - |
| Mouse Expression Set 430 | 1418283_PM_at | - | - |
| Murine Genome U74v2 Set | 101410_PM_at | - | - |
| Murine Genome U74v2 Set | 101410_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418283_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418283_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418283_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418283_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4920739 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004920739 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1223949 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1223949 | - | - |
| mouse-6 v1.1 (Illumina) | scl012740.2_183-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4920739 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl012740.2_183-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4920739 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1223949 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1223949 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl012740.2_183-S | - | - |

Cldn4
Genomic Map