Assay Details
| Gene Symbol: Cldn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009902.4 | NP_034032.1 | 1 | 951 | 85 | |
| GenBank mRNA | AB000715.1 | - | 1 | 132 | 85 | |
| AF095905.1 | - | 1 | 775 | 85 | ||
| AK002672.1 | - | 1 | 952 | 85 | ||
| AK006514.1 | - | 1 | 913 | 85 | ||
| AK012677.1 | - | 1 | 903 | 85 | ||
| AK165808.1 | - | 1 | 595 | 85 | ||
| BC023269.1 | - | 1 | 627 | 85 | ||
| BC032070.1 | - | 1 | 319 | 85 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cldn3
Genomic Map