Assay Details
| Gene Symbol: Gcn1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172719.2 | NP_766307.2 | 18 - 19 | 1880 | 60 | |
| GenBank mRNA | AK090137.1 | - | 18 - 19 | 1878 | 60 | |
| AK147257.1 | - | 18 - 19 | 1873 | 60 | ||
| AK154512.1 | - | 18 - 19 | 1873 | 60 | ||
| BC082548.1 | - | 5 - 6 | 658 | 60 | ||
| CD350892.1 | - | 5 - 6 | 658 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gcn1l1
Genomic Map