Assay Details
| Gene Symbol: Gcn1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172719.2 | NP_766307.2 | 26 - 27 | 3119 | 99 | |
| GenBank mRNA | AB093217.1 | - | 2 - 3 | 312 | 99 | |
| AK090137.1 | - | 26 - 27 | 3117 | 99 | ||
| AK147257.1 | - | 26 - 27 | 3112 | 99 | ||
| AK154512.1 | - | 26 - 27 | 3112 | 99 | ||
| BC082548.1 | - | 13 - 14 | 1897 | 99 | ||
| BC150735.1 | - | 26 - 27 | 3122 | 9087646 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gcn1l1
Genomic Map