Assay Details
| Gene Symbol: Gcn1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172719.2 | NP_766307.2 | 27 - 28 | 3192 | 71 | |
| GenBank mRNA | AB093217.1 | - | 3 - 4 | 385 | 71 | |
| AK089324.1 | - | 1 - 2 | 53 | 71 | ||
| AK090137.1 | - | 27 - 28 | 3190 | 71 | ||
| AK147257.1 | - | 27 - 28 | 3185 | 71 | ||
| AK154512.1 | - | 27 - 28 | 3185 | 71 | ||
| BC082548.1 | - | 14 - 15 | 1970 | 71 | ||
| BC150735.1 | - | 27 - 28 | 3195 | 9087646 | 71 | |
More Information
Set Membership: |
|

Gcn1l1
Genomic Map