Assay Details
| Gene Symbol: Gcn1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172719.2 | NP_766307.2 | 29 - 30 | 3467 | 64 | |
| GenBank mRNA | AB093217.1 | - | 5 - 6 | 660 | 64 | |
| AK089324.1 | - | 3 - 4 | 328 | 64 | ||
| AK090137.1 | - | 29 - 30 | 3465 | 64 | ||
| AK147257.1 | - | 29 - 30 | 3460 | 64 | ||
| BC082548.1 | - | 16 - 17 | 2245 | 64 | ||
| BC150735.1 | - | 29 - 30 | 3470 | 9087646 | 64 | |
More Information
Set Membership: |
|

Gcn1l1
Genomic Map