Assay Details
| Gene Symbol: Gcn1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172719.2 | NP_766307.2 | 31 - 32 | 3725 | 91 | |
| GenBank mRNA | AB093217.1 | - | 7 - 8 | 918 | 91 | |
| AK089324.1 | - | 5 - 6 | 586 | 91 | ||
| AK090137.1 | - | 31 - 32 | 3723 | 91 | ||
| AK147257.1 | - | 31 - 32 | 3718 | 91 | ||
| BC082548.1 | - | 18 - 19 | 2503 | 91 | ||
| BC150735.1 | - | 31 - 32 | 3728 | 9087646 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gcn1l1
Genomic Map