Assay Details
| Gene Symbol: Gcn1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172719.2 | NP_766307.2 | 34 - 35 | 4334 | 71 | |
| GenBank mRNA | AB093217.1 | - | 10 - 11 | 1527 | 71 | |
| AK089324.1 | - | 8 - 9 | 1195 | 71 | ||
| AK090137.1 | - | 34 - 35 | 4332 | 71 | ||
| AK147257.1 | - | 34 - 35 | 4327 | 71 | ||
| BC082548.1 | - | 21 - 22 | 3112 | 71 | ||
| BC150735.1 | - | 34 - 35 | 4337 | 9087646 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gcn1l1
Genomic Map