Assay Details
| Gene Symbol: Gcn1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172719.2 | NP_766307.2 | 36 - 37 | 4570 | 92 | |
| GenBank mRNA | AB093217.1 | - | 12 - 13 | 1763 | 92 | |
| AK090137.1 | - | 36 - 37 | 4568 | 92 | ||
| AK147257.1 | - | 36 - 37 | 4563 | 92 | ||
| BC056933.1 | - | 2 - 3 | 202 | 92 | ||
| BC082548.1 | - | 23 - 24 | 3348 | 92 | ||
| BC150735.1 | - | 36 - 37 | 4573 | 9087646 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gcn1l1
Genomic Map