Assay Details
| Gene Symbol: Gcn1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172719.2 | NP_766307.2 | 38 - 39 | 4968 | 69 | |
| GenBank mRNA | AB093217.1 | - | 14 - 15 | 2161 | 69 | |
| AK077013.1 | - | 2 - 3 | 290 | 69 | ||
| AK090137.1 | - | 38 - 39 | 4966 | 69 | ||
| AK147257.1 | - | 38 - 39 | 4961 | 69 | ||
| BC056933.1 | - | 4 - 5 | 600 | 69 | ||
| BC082548.1 | - | 25 - 26 | 3746 | 69 | ||
| BC150735.1 | - | 38 - 39 | 4971 | 9087646 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gcn1l1
Genomic Map