Assay Details
| Gene Symbol: Gcn1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172719.2 | NP_766307.2 | 42 - 43 | 5523 | 124 | |
| GenBank mRNA | AB093217.1 | - | 18 - 19 | 2716 | 124 | |
| AK077013.1 | - | 6 - 7 | 845 | 124 | ||
| AK090137.1 | - | 42 - 43 | 5520 | 124 | ||
| AK147257.1 | - | 42 - 43 | 5516 | 124 | ||
| BC056933.1 | - | 8 - 9 | 1155 | 124 | ||
| BC150735.1 | - | 42 - 43 | 5526 | 9087646 | 124 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gcn1l1
Genomic Map