Assay Details
| Gene Symbol: Gcn1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172719.2 | NP_766307.2 | 48 - 49 | 6419 | 75 | |
| GenBank mRNA | AB093217.1 | - | 24 - 25 | 3612 | 75 | |
| AK077013.1 | - | 12 - 13 | 1741 | 75 | ||
| AK090137.1 | - | 48 - 49 | 6416 | 75 | ||
| AK147257.1 | - | 48 - 49 | 6412 | 75 | ||
| BC038407.1 | - | 1 - 2 | 156 | 75 | ||
| BC056933.1 | - | 14 - 15 | 2051 | 75 | ||
| BC068244.1 | - | 4 - 5 | 673 | 75 | ||
| BC150735.1 | - | 48 - 49 | 6422 | 9087646 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gcn1l1
Genomic Map