Assay Details
| Gene Symbol: Gcn1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172719.2 | NP_766307.2 | 49 - 50 | 6689 | 58 | |
| GenBank mRNA | AB093217.1 | - | 25 - 26 | 3882 | 58 | |
| AK090137.1 | - | 49 - 50 | 6686 | 58 | ||
| AK147257.1 | - | 49 - 50 | 6682 | 58 | ||
| BC038407.1 | - | 2 - 3 | 426 | 58 | ||
| BC056933.1 | - | 15 - 16 | 2321 | 58 | ||
| BC068244.1 | - | 5 - 6 | 943 | 58 | ||
| BC150735.1 | - | 49 - 50 | 6692 | 9087646 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gcn1l1
Genomic Map