Assay Details
| Gene Symbol: Gcn1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172719.2 | NP_766307.2 | 50 - 51 | 6799 | 82 | |
| GenBank mRNA | AB093217.1 | - | 26 - 27 | 3992 | 82 | |
| AK077013.1 | - | 14 - 15 | 2118 | 82 | ||
| AK090137.1 | - | 50 - 51 | 6796 | 82 | ||
| AK147257.1 | - | 50 - 51 | 6792 | 82 | ||
| BC038407.1 | - | 3 - 4 | 536 | 82 | ||
| BC056933.1 | - | 16 - 17 | 2431 | 82 | ||
| BC068244.1 | - | 6 - 7 | 1053 | 82 | ||
| BC150735.1 | - | 50 - 51 | 6802 | 9087646 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gcn1l1
Genomic Map