Assay Details
| Gene Symbol: Gcn1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172719.2 | NP_766307.2 | 9 - 10 | 868 | 69 | |
| GenBank mRNA | AK041717.1 | - | 9 - 10 | 857 | 69 | |
| AK090137.1 | - | 9 - 10 | 866 | 69 | ||
| AK147257.1 | - | 9 - 10 | 861 | 69 | ||
| AK154512.1 | - | 9 - 10 | 861 | 69 | ||
| AK158040.1 | - | 9 - 10 | 859 | 69 | ||
| BC150735.1 | - | 9 - 10 | 871 | 9087646 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gcn1l1
Genomic Map