Assay Details
| Gene Symbol: Gcn1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172719.2 | NP_766307.2 | 10 - 11 | 945 | 75 | |
| GenBank mRNA | AK041717.1 | - | 10 - 11 | 934 | 75 | |
| AK090137.1 | - | 10 - 11 | 943 | 75 | ||
| AK147257.1 | - | 10 - 11 | 938 | 75 | ||
| AK154512.1 | - | 10 - 11 | 938 | 75 | ||
| AK157964.1 | - | 6 - 7 | 4806 | 75 | ||
| AK158040.1 | - | 10 - 11 | 936 | 75 | ||
| BC150735.1 | - | 10 - 11 | 948 | 9087646 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gcn1l1
Genomic Map