Assay Details
| Gene Symbol: Gcn1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172719.2 | NP_766307.2 | 13 - 14 | 1219 | 61 | |
| GenBank mRNA | AK037425.1 | - | 2 - 3 | 118 | 61 | |
| AK041717.1 | - | 13 - 14 | 1208 | 61 | ||
| AK090137.1 | - | 13 - 14 | 1217 | 61 | ||
| AK147257.1 | - | 13 - 14 | 1212 | 61 | ||
| AK154512.1 | - | 13 - 14 | 1212 | 61 | ||
| AK158040.1 | - | 13 - 14 | 1210 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gcn1l1
Genomic Map