Assay Details
| Gene Symbol: Stx1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016801.3 | NP_058081.2 | 4 - 5 | 295 | 74 | |
| GenBank mRNA | AK144399.1 | - | 1 - 2 | 185 | 74 | |
| BC047133.1 | - | 4 - 5 | 284 | 74 | ||
| BC057892.1 | - | 4 - 5 | 307 | 74 | ||
| BC100446.1 | - | 4 - 5 | 330 | 6595634 | 74 | |
| D45208.2 | - | 4 - 5 | 295 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stx1a
Genomic Map