Assay Details
| Gene Symbol: Lin54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115010.1 | NP_001108482.1 | 10 - 11 | 2010 | 68 | |
| NM_172714.4 | NP_766302.2 | 10 - 11 | 1878 | 68 | ||
| GenBank mRNA | AK029288.1 | - | 11 - 12 | 1324 | 68 | |
| AK030894.1 | - | 10 - 11 | 2012 | 68 | ||
| AK134320.1 | - | 10 - 11 | 1876 | 68 | ||
| AK220225.1 | - | 10 - 11 | 1873 | 68 | ||
| BC106126.1 | - | 9 - 10 | 1514 | 68 | ||
| BC125580.1 | - | 8 - 9 | 1005 | 68 | ||
| BC145025.1 | - | 8 - 9 | 1005 | 9053573 | 68 | |
More Information
Set Membership: |
|

Lin54
Genomic Map