Assay Details
| Gene Symbol: Oosp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037634.3 | NP_001032723.1 | 2 - 3 | 288 | 117 | |
| GenBank mRNA | BC099498.1 | - | 2 - 3 | 248 | 30931566 | 117 |
More Information
Set Membership: |
|

Oosp2
Genomic Map