Assay Details
| Gene Symbol: Camkk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199676.1 | NP_001186605.1 | 12 - 13 | 1456 | 68 | |
| NM_145358.2 | NP_663333.1 | 12 - 13 | 1456 | 68 | ||
| GenBank mRNA | AK031399.1 | - | 12 - 13 | 1381 | 68 | |
| AK032070.1 | - | 12 - 13 | 1429 | 68 | ||
| AK044660.1 | - | 12 - 13 | 1445 | 68 | ||
| AK122370.1 | - | 7 - 8 | 622 | 68 | ||
| BC023103.1 | - | 11 - 12 | 1289 | 3993474 | 68 | |
| BC025895.1 | - | 1 - 2 | 39 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Camkk2
Genomic Map