Assay Details
| Gene Symbol: Camkk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199676.1 | NP_001186605.1 | 15 - 16 | 1767 | 62 | |
| GenBank mRNA | AF453383.1 | - | 14 - 15 | 1550 | 62 | |
| AK032070.1 | - | 15 - 16 | 1740 | 62 | ||
| AK122370.1 | - | 10 - 11 | 933 | 62 | ||
| BC025895.1 | - | 3 - 4 | 221 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Camkk2
Genomic Map