Assay Details
| Gene Symbol: Adh6-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033581.1 | 2 - 3 | 193 | 123 | ||
| GenBank mRNA | AK004863.1 | - | 2 - 3 | 199 | 123 | |
| BY705068.1 | - | 2 - 3 | 194 | 123 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adh6-ps1
Genomic Map