Assay Details
| Gene Symbol: Chek2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016681.3 | NP_057890.1 | 1 - 2 | 27 | 60 | |
| GenBank mRNA | AK044913.1 | - | 1 - 2 | 28 | 60 | |
| AK170235.1 | - | 1 - 2 | 62 | 60 | ||
| BC056617.1 | - | 1 - 2 | 51 | 4977960 | 60 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK044913.1, BC056617.1, NM_016681.3 transcripts.
Set Membership: |
|

Chek2
Genomic Map