Assay Details
| Gene Symbol: Chek2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016681.3 | NP_057890.1 | 6 - 7 | 848 | 154 | |
| GenBank mRNA | AF086905.1 | - | 5 - 6 | 809 | 154 | |
| AK044913.1 | - | 6 - 7 | 849 | 154 | ||
| AK170235.1 | - | 5 - 6 | 792 | 154 | ||
| BC056617.1 | - | 6 - 7 | 872 | 4977960 | 154 | |
More Information
Updated Mapping Information
Set Membership: |
|

Chek2
Genomic Map