Assay Details
| Gene Symbol: Fbrsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142642.1 | NP_001136114.1 | 11 - 12 | 1929 | 64 | |
| NM_028596.2 | NP_082872.2 | 5 - 6 | 1045 | 64 | ||
| GenBank mRNA | AK031967.1 | - | 5 - 6 | 1015 | 64 | |
| BC059932.1 | - | 9 - 10 | 2715 | 64 | ||
| BC151196.1 | - | 6 - 7 | 938 | 9088084 | 64 | |
| BU614458.1 | - | 2 - 3 | 237 | 64 | ||
More Information
Set Membership: |
|

Fbrsl1
Genomic Map