Assay Details
| Gene Symbol: Fbrsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142642.1 | NP_001136114.1 | 15 - 16 | 2229 | 55 | |
| NM_028596.2 | NP_082872.2 | 8 - 9 | 1273 | 55 | ||
| GenBank mRNA | AK031967.1 | - | 8 - 9 | 1243 | 55 | |
| BC059932.1 | - | 13 - 14 | 3015 | 55 | ||
| BC069994.1 | - | 3 - 4 | 230 | 55 | ||
| BC151196.1 | - | 10 - 11 | 1238 | 9088084 | 55 | |
| BU614458.1 | - | 5 - 6 | 465 | 55 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fbrsl1
Genomic Map