Assay Details
| Gene Symbol: Fbrsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142642.1 | NP_001136114.1 | 1 - 2 | 621 | 73 | |
| GenBank mRNA | BC059932.1 | - | 1 - 2 | 604 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbrsl1
Genomic Map