Assay Details
| Gene Symbol: Fbrsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142642.1 | NP_001136114.1 | 3 - 4 | 937 | 76 | |
| GenBank mRNA | BC059932.1 | - | 3 - 4 | 920 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbrsl1
Genomic Map